Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GPR37 (Human) Recombinant Protein

Catalog No. 89998444 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-444 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-444 Supplier Abnova™ Supplier No. H00002861G01
Add to Cart
Add to Cart

Human GPR37 full-length ORF (NP_005293.1) recombinant protein without tag.

  • Sequence: MRAPGALLARMSRLLLLLLLKVSASSALGVAPASRNETCLGESCAPTVIQRRGRDAWGPGNSARDVLRARAPREEQGAAFLAGPSWDLPAAPGRDPAAGRGAEASAAGPPGPPTRPPGPWRWKGARGQEPSETLGRGNPTALQLFLQISEEEEKGPRGAGISGRSQEQSVKTVPGASDLFYWPRRAGKLQGSHHKPLSKTANGLAGHEGWTIALPGRALAQNGSLGEGIHEPGGPRRGNSTNRRVRLKNPFYPLTQESYGAYAVMCLSVVIFGTGIIGNLAVMCIVCHNYYMRSISNSLLANLAFWDFLIIFFCLPLVIFHELTKKWLLEDFSCKIVPYIEVASLGVTTFTLCALCIDRFRAATNVQMYYEMIENCSSTTAKLAVIWVGALLLALPEVVLRQLSKEDLGFSGRAPAERCIIKISPDLPDTIYVLALTYDSARLWWYFGCYFCLPTLFTITCSLVTARKIRKAEKACTRGNKRQIQLESQMNCTVVALTILYGFCIIPENICNIVTAYMATGVSQQTMDLLNIISQFLLFFKSCVTPVLLFCLCKPFSRAFMECCCCCCEECIQKSSTVTSDDNDNEYTTELELSPFSTIRREMSTFASVGTHC

Specifications

Accession Number NP_005293.1
Gene ID (Entrez) 2861
Name G protein-coupled receptor 37 (endothelin receptor type B-like)
Preparation Method Wheat germ expression system
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias EDNRBL, PAELR, hET(B)R-LP
Gene Symbol GPR37
Species Wheat Germ (in vitro)
Buffer 25 mM Tris-HCl of pH 8.0 containing 2% glycerol
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.