Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR37L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106838 Supplier Invitrogen™ Supplier No. PA5106838
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ETBR2.

This polyclonal antibody was raised in Rabbit against GPR37L1. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPR37L1. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR37L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR37L1
Gene Accession No. O60883, Q99JG2, Q9QYC5
Gene Alias AW047233; CAG-18; D0Kist8; Endothelin B receptor-like protein 2; endothelin B receptor-like protein-2; endothelin type b receptor-like protein 2; ET(B)R-LP-2; ETBRLP2; ETBR-LP-2; G protein-coupled receptor 37 like 1; G protein-coupled receptor 37-like 1; G protein-coupled receptor 37-lke 1; GPCR/CNS2; GPR37L1; G-protein coupled receptor 37 like 1; G-protein coupled receptor 37-like 1; orphan Gpr37-like protein 1; prosaposin receptor GPR37L1
Gene Symbols GPR37L1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR37L1(Accession O60883), corresponding to amino acid residues A8-Q58.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171469, 252939, 9283
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.