Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115321 Supplier Invitrogen™ Supplier No. PA5115321
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR40.

This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR40
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FFAR1
Gene Accession No. O14842, Q76JU9, Q8K3T4
Gene Alias FFA1R; FFAR; Ffar1; Free fatty acid receptor 1; G protein-coupled receptor 40; G protein-coupled receptor GPR40; GPCR40; GPR40; G-protein coupled receptor 40; HGNC:4498
Gene Symbols FFAR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FFAR1(Accession O14842), corresponding to amino acid residues F191-N241.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233081, 266607, 2864
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.