Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550978 Supplier Invitrogen™ Supplier No. PA550978
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR50 Polyclonal Antibody detects endogenous levels of GPR50 protein.

Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR50
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR50
Gene Accession No. Q13585
Gene Alias G protein-coupled receptor 50; Gpr50; G-protein-coupled receptor 50; H9; Mel1c; melatonin-related receptor; MTNRL
Gene Symbols GPR50
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human GPR50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9248
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.