Learn More
GPR56 Mouse anti-Human, Mouse, PE, Clone: CG4.rMAb (also known as CG4), BD Pharmingen™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | GPR56 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CG4.rMAb (also known as CG4) |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. |
| Gene Alias | G protein-coupled receptor 56; ADGRG1; BFPP; BPPR; TM7LN4; TM7XN1; testicular tissue protein Li 77 |
| Host Species | Mouse |
| Immunogen | Human GPR56 Recombinant Protein |
| Quantity | 100 Tests |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.