Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR68 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549323
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549323 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549323 Supplier Invitrogen™ Supplier No. PA549323
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with bovine and human based on sequence homology.

GPR68 (also known as OGR1 - ovarian cancer G-protein coupled receptor 1) is a proton-sensing receptor involved in pH homeostasis. It represents an osteoblastic pH sensor regulating the cell-mediated response to acidosis in bone. GPR68 mediates its action by association with G proteins that stimulate inositol phosphate (IP) production or CA2+ mobilization. The receptor is almost silent at pH 7.8 but fully activated at pH 6.8. Also found to serve as a tumor suppressor in prostate cancer. Mutations to the gene result in amelogenesis imperfecta, hypomaturation type 2A6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR68
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GPR68
Gene Accession No. Q8BFQ3
Gene Alias BB131428; Brgr1; Brgrb; G protein-coupled receptor 68; GPR12A; Gpr68; G-protein coupled receptor 68; Ogr1; OGR-1; ovarian cancer G protein-coupled receptor, 1; ovarian cancer G-protein coupled receptor 1; Sphingosylphosphorylcholine receptor
Gene Symbols GPR68
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 200-225 amino acids from the Central region of mouse Gpr68.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238377
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.