Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533765
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533765 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533765 Supplier Invitrogen™ Supplier No. PA533765
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Rat, Dog, Bat, Hamster, Panda, Rabbit, Opossum (100%).

The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR81
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene HCAR1
Gene Accession No. Q9BXC0
Gene Alias FKSG80; G protein-coupled receptor 104; G protein-coupled receptor 81; GPR104; GPR81; G-protein coupled receptor 104; G-protein coupled receptor 81; HCA1; HCAR1; hydrocarboxylic acid receptor 1; hydroxycarboxylic acid receptor 1; hydroxy-carboxylic acid receptor 1; LACR1; lactate receptor 1; TAGPCR; TA-GPCR; T-cell activation G protein-coupled receptor
Gene Symbols HCAR1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 2nd extracellular domain of human GPR81.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100345808, 100766347, 27198, 369158, 486252
Target Species Human, Rat, Canine, Rabbit, Monkey, Hamster, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.