Learn More
Description
IHC(P) analysis shows positive staining of GPR82 in human kidney tubules and germinal center of human lymph node. WB analysis shows GPR82 in glyco-protein enriched fractions from mock transfected or GPR82 overexpressed 293 cells. GPR82 exists in different glycosylated forms which may cause the molecular weight detected by WB to vary.
Specifications
Specifications
| Antigen | GPR82 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.05% sodium azide |
| Gene | GPR82 |
| Gene Accession No. | Q96P67 |
| Gene Alias | G protein-coupled receptor 82; GPR82; Probable G-protein coupled receptor 82 |
| Gene Symbols | GPR82 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
