Learn More
GPR85 Polyclonal Antibody, Invitrogen™
Description
PA1-24547 detects G Protein-Coupled Receptor GPR85/SREB2 in Human samples.
Specifications
Specifications
| Antigen | GPR85 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | GPR85 |
| Gene Accession No. | P60893 |
| Gene Alias | 2900026B03Rik; FLJ50993; FLJ51159; FLJ99433; G protein-coupled receptor 85; Gpr85; PKrCx1; probable G-protein coupled receptor 85; Rgpr85; seven transmembrane helix receptor; SREB; SREB2; Srep2; Super conserved receptor expressed in brain 2 |
| Gene Symbols | GPR85 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.