Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR87 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3043 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3043 Supplier Invitrogen™ Supplier No. PA3043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IHC(P) analysis shows positive staining of GPR87 in human kidney tubules. WB analysis shows GPR87 in glyco-protein enriched fractions from GPR84 or GPR87 overexpressed 293 cells. The GPR87 antiserum showed specific detection of GPR87 over other tested GPRs. GPR87 exists in different glycosylated forms.

G protein-coupled receptors play a role in cell communication. They are characterized by an extracellular N terminus, 7 transmembrane regions, and an intracellular C terminus. GPR87 has been reported to be expressed in human prostate and rat stomach and fetal tissues. ESTs have been isolated from B-cell/lung/testis, breast, heart, jaw, prostate, and testis libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR87
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene GPR87
Gene Accession No. Q9BY21
Gene Alias FKSG78; G protein-coupled receptor 87; G protein-coupled receptor 95; GPR87; GPR95; G-protein coupled receptor 87; G-protein coupled receptor 95; KPG_002; MGC131898; orphan GPCR 87
Gene Symbols GPR87
Host Species Rabbit
Immunogen KLH conjugated COOH-terminal amino acids of 339-358 (RSLQSVRRSEVRIYYDYTDV).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53836
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.