Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR91 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599450 Supplier Invitrogen™ Supplier No. PA599450
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR91.

This polyclonal antibody was raised in Rabbit against GPR91. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPR91. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR91
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SUCNR1
Gene Accession No. Q6IYF9, Q99MT6, Q9BXA5
Gene Alias G protein-coupled receptor 91; GPR91; G-protein coupled receptor 91; G-protein-coupled receptor 91; p2u2; P2Y purinoceptor 1; P2Y purinoceptor 1-like; Succinate receptor; Succinate receptor 1; SUCNR1; SUCR1
Gene Symbols SUCNR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SUCNR1(Accession Q9BXA5), corresponding to amino acid residues T108-L158.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 408199, 56670, 84112
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.