Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3049 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3049 Supplier Invitrogen™ Supplier No. PA3049
  • $601.52
    $481.65 / Each of 1

    Save $119.87

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

WB analysis shows GPR97 in glyco-protein enriched fractions from mock transfected, GPR88 or GPR57 overexpressed 293 cells. GPR97 exists in different glycosylated forms.

GPR97 is expressed exclusively in pre-B and pre-T cells in the thymus but not in mature lymphoid or non-lymphoid tissues. ESTs have been isolated from spleen libraries. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Monkey (94%) Mouse, Rat (89%) Rabbit (83%)..
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR97
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene ADGRG3
Gene Accession No. Q86Y34
Gene Alias ADGRG3; Adhesion G protein-coupled receptor G3; G protein-coupled receptor 97; GPR97; G-protein coupled receptor 97; G-protein coupled receptor PGR26; PB99; PGR26; probable G-protein coupled receptor 97
Gene Symbols ADGRG3
Host Species Rabbit
Immunogen KLH conjugated COOH-terminal amino acids of 529 to 549 (QSTTVSSSTARLDQAHSASQE).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222487
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.