Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPRC5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532853
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532853 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532853 Supplier Invitrogen™ Supplier No. PA532853
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%); Mouse, Rat, Dog, Bovine, Elephant, Panda, Horse, Rabbit, Pig, Opossum, Turkey, Chicken, Xenopus, Pufferfish, Zebrafish, Stickleback (94%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

This gene encodes a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The encoded protein may modulate insulin secretion and increased protein expression is associated with type 2 diabetes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPRC5B
Gene Accession No. Q9NZH0
Gene Alias A-69G12.1; G protein-coupled receptor class C group 5 member B; G protein-coupled receptor, class C, group 5, member B; G protein-coupled receptor, family C, group 1, member B; GPRC5B; G-protein coupled receptor family C group 5 member B; RAIG2; RAIG-2; retinoic acid responsive gene protein; retinoic acid-induced gene 2 protein
Gene Symbols GPRC5B
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GPRC5B.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51704
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.