Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109786 Supplier Invitrogen™ Supplier No. PA5109786
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSKAESTPEI AEQRGERDSQ TQAILTKLKC AAGLAELAAR KYKQAAKCLL LASFDHCDFP ELLSPSNVAI YGGLCALATF DRQELQRNVI SSSSFKLFLE LEPQVRDIIF KFYESKYASC LKMLDEMKDN LLLDMYLAPH VRTLYTQIRN RALIQYFSPY VSADMHRMAA AFNTTVAALE DELTQLILEG LISARVDSHS KILYARDVDQ RSTTFEKSLL MGKEFQRRAK AMMLRAAVLR NQIHVKSPPR EGSQGELTPA NSQSRMSTNM.

OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPS1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GPS1
Gene Accession No. P97834, Q13098, Q99LD4
Gene Alias COP9 signalosome complex subunit 1; Cops1; CSN1; G protein pathway suppressor 1; Gps1; GPS-1; JAB1-containing signalosome subunit 1; MFH; MFH protein; MGC71287; Protein MFH; R75577; SGN1; signalosome subunit 1
Gene Symbols GPS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 258-527 of human GPS1 (NP_997657.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117039, 209318, 2873
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.