Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

X3 Industrial Grade Vinyl Gloves
GREENER_CHOICE

Catalog No. 19227024
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-227-024 Medium
19-227-023 Small
19-227-026 X-Large
19-227-025 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-227-024 Supplier X3 Supplier No. GPX344100
Add to Cart
Add to Cart
This item is not returnable. View return policy

GPX3™ clear, vinyl industrial grade gloves deliver comfort, dexterity, and tactile sensitivity. Dependable and effective barrier protection for lightweight tasks.

  • Constructed from lightweight, 3 mil vinyl
  • Tactile sensitivity, comfort, and dexterity at a lower price than nitrile or latex
  • Vinyl is a synthetic material made from polyvinyl chloride with a looser fit and smooth, comfortable feel
  • Dependable barrier protection against common cleaning solutions, hair dyes, and other non-hazardous materials
  • Suited for work environments such as food service, painting, janitorial, sanitation, plumbing, salon & beauty
  • Ideal for light-industrial applications where frequent glove changes are required
  • Latex-free for those allergic to natural rubber
  • Powder free to prevent messy residue

Specifications

Applications Industrial
AQL 4
Color Clear
Disposable Single-use
Finger Thickness (English) 3 mil
Finger Thickness (Metric) 0.08 mm
Hand Ambidextrous
Latex Free Yes
Material Vinyl
Palm Thickness (English) 3 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Size Medium
Series GPX3
Length (English) 9.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.