Learn More
Invitrogen™ GPX4 Recombinant Rabbit Monoclonal Antibody (7O4Q9)

Description
GPX4 Recombinant Monoclonal Antibody for ELISA, Western Blot
Specifications
Specifications
| Antigen | GPX4 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 7O4Q9 |
| Concentration | 0.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.09% sodium azide; pH 7.3 |
| Gene | GPX4 |
| Gene Accession No. | O70325, P36969, P36970 |
| Gene Alias | glutathione peroxidase 4; GP-GPx4; Gpx4; gpx-4; GSHPx-4; HGNC:4556; MCSP; mtPHGPx; Phgpx; phospholipid hydroperoxidase; phospholipid hydroperoxide glutathione peroxidase; phospholipid hydroperoxide glutathione peroxidase, mitochondrial; phospholipid hydroperoxide glutathione peroxidase, nuclear; SMDS; snGPx; snPHGPx; sperm nuclei glutathione peroxidase; sperm nucleus glutathione peroxidase |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.