Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPX7 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728788 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8788 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8788 Supplier Proteintech Group Inc Supplier No. 135011AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.

Specifications

Antigen GPX7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GPX7
Gene Accession No. Q96SL4
Gene Alias CL683, glutathione peroxidase 7, GPx 7, GPX6, GPX7, GSHPx 7
Gene Symbols GPX7
Host Species Rabbit
Immunogen GPX7 Fusion Protein Ag4328
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2882
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.