Learn More
Sigma Aldrich Fine Chemicals Biosciences Grace Bio Labs PATHR prote
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GBL80502520EA
Encompass
The Grace Bio-Labs PATH protein microarray slides are a non-porous nitrocellulose film slides. BenefitsLow and consistant fluorescent backgroundsHigher protein bindingSuperior signal-to-noise
Catalog No. 50-290-8683