Learn More
United Scientific™ Graduated Cylinder, Double Scale, With Stopper
SDPSCI_ED
Catalog No. S15953 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Catalog No. Capacity (Metric)
S15953 250 mL
1 options
Catalog No. S15953 Supplier United Scientific™ Supplier No. CY2981250
Add to Cart
Add to Cart
Cylinders are made from borosilicate glass and ideal for everyday lab use.
  • Cylinders comply with ASTM E1272, Class A standards, batch certified.
  • Each cylinder includes a hollow glass stopper.
  • Double metric scale, calibrated to contain.
  • Borosilicate glass cylinders feature heavy uniform wall tubing and strong, stable, hexagonal bases.
  • All cylinders come with bumper guards.
Specifications
Material Borosilicate Glass (3.3)
Capacity (Metric) 250 mL
Graduations 2 mL
Class Class A
Tolerance ASTM E1272
Product Type Graduated Cylinder