Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Graduated Cylinders, Double Scale, Borosilicate Glass, Class B
SDPSCI_ED

Catalog No. S15814 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.5 mL
1 mL
5 mL
10 mL
20 mL
Catalog No. Capacity (Metric) Graduations
S15814 100 mL 1 mL
S16051 5 mL 0.5 mL
S15996 10 mL 0.5 mL
S16018 25 mL 0.5 mL
S16090 50 mL 1 mL
S16050 250 mL 10 mL
S15813 500 mL 5 mL
S16091 1000 mL 10 mL
S16068 2000 mL 20 mL
9 options

Catalog No. S15814

Supplier: United Scientific™ UNCYLNGB100

Only null left
Add to Cart
Add to Cart

Cylinders are made from borosilicate glass and ideal for everyday lab use.

  • United Scientific™ graduated cylinders feature a double metric scale, calibrated to contain.
  • Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing and attached glass hexagonal bases.
  • Made from heavy-duty borosilicate 3.3 glass
  • Meets ASTM E1272 Class B tolerances for precision and are individually calibrated to contain
  • One-piece molded glass construction with hexagonal base and adjustable plastic bumper guard
  • Low temperature gradient, can withstand high temperatures and thermal shocks
  • All cylinders include bumper guards.

Specifications

Type Graduated Cylinder
Class B
Tolerance Metric ASTM E1272
Material Borosilicate Glass (3.3)
Capacity (Metric) 100 mL
Graduations 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.