Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Pulp/Aluminum Caps

Catalog No. 02991772
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
480 mL
Closure Size:
33-400 mm
38-400 mm
48-400 mm
58-400 mm
70-400 mm
Packaging:
Bulk Pack
Quantity:
144/Cs.
288/Cs.
432/Cs.
48/Cs.
96/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size Packaging Quantity
02-991-772 60 mL 38-400 mm Bulk Pack 288/Cs.
02-992-226 120 mL 48-400 mm Bulk Pack 144/Cs.
02-991-790 480 mL 70-400 mm Bulk Pack 48/Cs.
02-991-856 30 mL 33-400 mm Bulk Pack 432/Cs.
02-991-737 240 mL 58-400 mm Bulk Pack 96/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-991-772 Supplier Qorpak™ Supplier No. GLC01482

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For mixing, measuring and storage

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Specifications

Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Diameter (English) 1.65 in.
Material Glass
Packaging Bulk Pack
Quantity 288/Cs.
Closure Color Black
Height (Metric) 83 mm
Height (English) 3.27 in.
Closure Liner Pulp/Aluminum Foil
Closure Material Phenolic
Closure Size 38-400 mm
Diameter (Metric) 42 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.