Learn More
Bio Plas Graduated Screw Cap Tube with Conical Base and Writing Area
Catalog No. 22025055
Change view
Click to view available options
Capacity (Metric):
1.5 mL
Catalog No. Capacity (Metric)
22-025-055 1.5 mL
1 options
Catalog No. 22-025-055 Supplier Bio Plas Supplier No. 4202G
Add to Cart
Add to Cart
Screw cap tube with conical base and writing area.
  • 1.5 mL graduated screw cap tube
  • Conical base
  • Writing area
  • Clear
Specifications
Cap Style Screw Cap
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Translucent
Material Polypropylene (Medical Grade)
No. per Pack 500/Pk.
Volume (Metric) 1.5 mL
Sterility Non-sterile
Graduated Yes
Closure Type Not Included
Fit for Purpose Certified Quality
Bottom Shape Conical
Max. RCF 20000 x g
Shape Cylindrical
Product Type Screw-cap Microtube