Learn More
United Scientific Supplies Graham Condensers, 24/40 Joint
SDPSCI_ED
Catalog No. S43785 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Size:
200 mm
300 mm
400 mm
500 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S43785 300 mm
S43784 200 mm
S43786 400 mm
S43787 500 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S43785 Supplier United Scientific Supplies Supplier No. CNG300
Only null left
Add to Cart
Add to Cart
Designed for use in distillation applications
  • Designed for use in distillation applications
  • Constructed of borosilicate glass
  • A coiled inner tube provides additional surface area for highly efficient cooling
  • Socket is 24/40 with a screw thread connector
Specifications
Connection Type Screw Thread
Material Borosilicate Glass
Type Graham Condenser
For Use With (Application) Distillation
For Use With (Equipment) Distillation Glassware
Rotor System Spiral
Size 300 mm