Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graham-Field™ Stethoscopes
SDP

Catalog No. 18999438A
Change view
Click to view available options
Product Type:
Dual Head Stethoscope
Single Head Stethoscope
Sprague Rappaport Stethoscope
Color:
Black
Blue
Gray
Green
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
18-999-438A Dual Head Stethoscope Black
19-065891 Single Head Stethoscope Black
19-027456 Single Head Stethoscope Gray
19-027455 Single Head Stethoscope Blue
19-027459 Dual Head Stethoscope Blue
19-027466 Sprague Rappaport Stethoscope Black
19-027461 Dual Head Stethoscope Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 18-999-438A Supplier Graham-Field™ Supplier No. 400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Single Head Stethoscope

  • Anodized aluminum chestpiece with finger rest
  • Adjustable chrome-plated brass binaurals
  • 22 in. (56cm) latex-free “Y” tubing
Double Head Stethoscope
  • Rotating anodized aluminum chestpiece with recessed non-chill ring
  • Adjustable chrome-plated brass binaurals
  • 22 in. (56cm) latex-free “Y” tubing
Sprague Rappaport-Type Stethoscope
  • Versatile enough for infants, children and adults
  • Delivers accurate detection of high, medium and low frequency sounds
  • Comes complete with accessory pouch including three interchangeable bells, adult diaphragm, pediatric diaphragm, and two pairs of eartips
  • 22 in. (56cm) latex-free tubing

Specifications

Product Type Dual Head Stethoscope
Color Black
For Use With (Application) To detect high, medium and low frequency sounds for infants, children and adults
Includes Anodized aluminum rotating chestpiece with recessed non-chill ring, adjustable chrome plated brass binaurals, 22 in. latex-free Y-tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.