Learn More
Description
Gram Stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.
- Complete stain kits reduce the mess associated with Gram staining procedures and include a compact tray for holding the 250mL reagent bottles
- Break-resistant bottles features no-drip, keep-clean cap spouts that make regulating the flow easy and that help to prevent spots and spills
- Color-coded labels allow quick identification of the reagent
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 3.8 L |
| Certifications/Compliance | CE |
| Format | Liquid |
| For Use With (Application) | Used to stain microorganisms from cultures or specimens by the differential gram method |
| Color | Purple |
| Storage Requirements | 15° to 30°C |
| Stain Type | Crystal Violet |
| Product Type | Crystal Violet |
| Packaging | Bulk storage container |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
