Learn More
Description
Gram Staining is used to differentiate gram-positive and gram-negative microorganisms from cultures or specimens. This Gram Decolorizer is an acetone (25%)/isopropanol (75%) solution for rapid decolorization, prior to counterstaining.
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded label allows quick identification of reagent
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 4 x 250mL |
| Certifications/Compliance | CE |
| Format | Liquid |
| For Use With (Application) | Staining microorganisms from cultures or specimens by the differential Gram method. |
| Color | Clear ad colorless |
| Closure Type | No drip flip cap |
| Composition | Acetone (75%), Isopropanol (75%) |
| Storage Requirements | 15° to 30°C |
| Stain Type | Decolorizer |
| Product Type | Decolorizer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
