Learn More
Invitrogen™ Gram Negative Endotoxin Monoclonal Antibody (GNE11-222.4), Invitrogen™

Description
MA1-10689 detects Endotoxin from Sh. sonnei, S. typhimurium, S. marcescens, P. mirabilis, P. vulgaris, and A. calcoaceticus samples. MA1-10689 has been successfully used in ELISA applications. The MA1-10689 immunogen is E. coli serotypes: O:111, B4 J5.
Specifications
Specifications
| Antigen | Gram Negative Endotoxin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | GNE11-222.4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Host Species | Mouse |
| Immunogen | E. coli serotypes: O:111, B4 J5. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.