Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gram Negative Endotoxin Monoclonal Antibody (HEC11-445.2), Invitrogen™
GREENER_CHOICE

Catalog No. 01671922
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-922 500 μg
PIMA546162 100 μg
PIMA546091 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-922 Supplier Invitrogen™ Supplier No. MA110691
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10691 detects Endotoxin from E. aerogenes, S. marcescens, P. mirabilis, A. calcoaceticus, and Ps. aeruginosa samples. MA1-10691 has been successfully used in ELISA applications. The MA1-10691 immunogen is E. coli serotypes: O:111, B4 J5.

Endotoxins are part of the outer membrane of the cell wall of gram-negative bacteria. Endotoxins are invariably associated with gram-negative bacteria whether the organisms are pathogens or not. Although the term endotoxin is occasionally used to refer to any cell-associated bacterial toxin, it is properly reserved to refer to the lipopolysaccharide complex associated with the outer membrane of gram-negative bacteria such as E. coli, Salmonella, Shigella, Pseudomonas, Neisseria, Haemophilus, and other leading pathogens. The biological activity of endotoxin is associated with the lipopolysaccharide (LPS). Toxicity is associated with the lipid component (Lipid A) and immunogenicity is associated with the polysaccharide components. The cell wall antigens (O antigens) of gram-negative bacteria are components of LPS. LPS elicits a variety of inflammatory responses in an animal. Because it activates complement by the alternative (properdin) pathway, it is often part of the pathology of gram-negative bacterial infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gram Negative Endotoxin
Applications ELISA
Classification Monoclonal
Clone HEC11-445.2
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen E. coli serotypes: O:111, B4 J5.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.