Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gram Positive Bacteria LTA Monoclonal Antibody (G43J)
GREENER_CHOICE

Catalog No. MA17402 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17402 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17402 Supplier Invitrogen™ Supplier No. MA17402
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7402 antibody reacts with Gram positive bacteria LTA in bacterial and infected tissue samples. MA1-7402 has been successfully used in ELISA, Western blot, and immunofluorescence applications. Suggested, but not tested in immunohistochemistry applications. The MA1-7402 antibody was raised against Lipoteichoic acid consistent with that of gram positive bacteria.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Gram Positive Bacteria LTA
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone G43J
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Lipoteichoic acid
Host Species Mouse
Immunogen Lipoteichoic acid consistent with that of gram pos. bacteria.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.