Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Biochemical Reagents
- Staining Reagents
- Stains and Dyes
- ENG Scientific Gram Stain Kit
ENG Scientific Gram Stain Kit
Catalog No. ES6151
Change view
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
2 x 950 mL
4 x 250 mL
Packaging:
Gallon Bottle
Kit (Bottles with flip-top cap)
Pack (Bottles with flip-top cap)
Slim-line Bottle
Product Type:
Acetone-Alcohol
Acetone-Alcohol, Carbol Fuchsin
Basic Fuchsin
Crystal Violet
Ethanol 70%
Ethanol 70%, Basic Fuchsin
Ethanol 70%, Carbol Fuchsin
Ethanol 95%
Ethanol 95%, Basic Fuchsin
Ethanol 95%, Carbol Fuchsin
Gram Iodine
Safranin
20 product options available for selection
| Catalog No. | Quantity | Packaging | Product Type |
|---|---|---|---|
| ES6151 | 1 x 3800 mL (1 gal.) | Slim-line Bottle | Crystal Violet |
| ES6050 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Ethanol 95% |
| ES6281 | 1 x 3800 mL (1 gal.) | Slim-line Bottle | Acetone-Alcohol |
| ES6000 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Ethanol 70% |
| ES6090 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Acetone-Alcohol |
| ES6097 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Acetone-Alcohol, Carbol Fuchsin |
| ES6096 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Ethanol 95%, Carbol Fuchsin |
| ES6400 | 4 x 250 mL | Pack (Bottles with flip-top cap) | Safranin |
| ES6200 | 4 x 250 mL | Pack (Bottles with flip-top cap) | Gram Iodine |
| ES6451 | 1 x 3800 mL (1 gal.) | Slim-line Bottle | Safranin |
| ES6100 | 4 x 250 mL | Pack (Bottles with flip-top cap) | Crystal Violet |
| ES6095 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Ethanol 70%, Carbol Fuchsin |
| ES6080 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Ethanol 95%, Basic Fuchsin |
| ES6070 | 4 x 250 mL | Kit (Bottles with flip-top cap) | Ethanol 70%, Basic Fuchsin |
| ES6320 | 2 x 950 mL | Pack (Bottles with flip-top cap) | Ethanol 70% |
| ES6455 | 4 x 250 mL | Pack (Bottles with flip-top cap) | Basic Fuchsin |
| ES6280 | 1 x 3800 mL (1 gal.) | Gallon Bottle | Acetone-Alcohol |
| ES6350 | 1 x 3800 mL (1 gal.) | Gallon Bottle | Ethanol 70% |
| ES6260 | 4 x 250 mL | Pack (Bottles with flip-top cap) | Acetone-Alcohol |
| ES6251 | 1 x 3800 mL (1 gal.) | Slim-line Bottle | Gram Iodine |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
Description
- Available in various counterstains
- Various decolorizers available to suit individual technique preference
- Flip-top cap on 250 mL and 950 mL sizes
- In vitro diagnostic use
Specifications
Specifications
| Quantity | 1 x 3800 mL (1 gal.) |
| For Use With (Application) | In vitro diagnostic use |
| Product Type | Crystal Violet |
| Packaging | Slim-line Bottle |
For in vitro diagnostic use only.
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?