Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gramine 98.0+%, TCI America™
SDP

Catalog No. D0653250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0653250G 250 g
D065325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0653250G Supplier TCI America Supplier No. D0653250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87-52-5
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD00005629
InChI Key OCDGBSUVYYVKQZ-UHFFFAOYSA-N
Synonym gramine, 3-dimethylaminomethyl indole, donaxine, donaxin, gramin, 1h-indole-3-methanamine, n,n-dimethyl, beta-dimethylaminomethylindole, 1-1h-indol-3-yl-n,n-dimethylmethanamine, indol-3-ylmethyldimethylamine, 1h-indol-3-ylmethyl dimethylamine
PubChem CID 6890
ChEBI CHEBI:28948
IUPAC Name 1-(1H-indol-3-yl)-N,N-dimethylmethanamine
SMILES CN(C)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 132°C
Quantity 250 g
Formula Weight 174.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Gramine
RTECSNumber NL7525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.