Learn More
Thermo Scientific GRAMS/3D Spectroscopy Software
Supplier:  Thermo Scientific INF15003
Encompass
Manipulate large 3D data sets in real-time on their PCs and see unseen information hidden in multidimensional data with Thermo Scientific GRAMS/3D Spectroscopy Software, an interactive visualization package for spectroscopy data and chemometric modeling.
Catalog No. NC1595323
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.