Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granulocyte Marker Monoclonal Antibody (HIS48), eBioscience™
GREENER_CHOICE

Catalog No. 5012663
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-126-63 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-126-63 Supplier Invitrogen™ Supplier No. 14057082
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

The HIS48 monoclonal antibody reacts with a molecule expressed on the surface of all rat granulocytes as well as monocytes. The antigen recognized by HIS48 is also expressed by erythroid lineage cells during their developmental stages in the bone marrow. HIS48 is commonly used as a phenotypic marker for rat granulocytes when used in combination with other markers (such as CD11b in which granulocytes are CD11b high/Rat Granulocyte (His48) medium expressers) Applications Reported: The HIS48 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. Applications Tested: The HIS48 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Granulocytes are a category of white blood cells characterized by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulocyte Marker
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HIS48
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF3; CSF3OS; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; Granulocyte Marker; granulocyte-colony stimulating factor; lenograstim; Pluripoietin; PML; PMN
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.