Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granulysin Monoclonal Antibody (eBioDH2 (DH2)), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5011218
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-18 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-18 Supplier Invitrogen™ Supplier No. 12882842
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The eBioDH2 monoclonal antibody reacts with human granulysin, a protein involved in the cytolytic pathway. Granulysin is a member of the saposin-like protein (SAPLIP) family. Two proteins 9 and 15kDa are thought to be post translation modified versions of the 15kDa protein with a change in confirmation. Granulysin is found in NK and activated T cells and the expression is confined to the cytolytic granules which also contains perforin and granzyme. Granulysin with perforn are suspected to function synergistically to kill bacteria and induce apoptosis in nucleated cells. Applications Reported: This eBioDH2 (DH2) antibody has been reported for use in intracellular staining followed by flow cytometric analysis and immunocytochemistry. Applications Tested: This eBioDH2 (DH2) antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

GNLY is a member of the saposin-like protein (SAPLIP) family and is located in the cytotoxic granules of T cells, which are released upon antigen stimulation. GNLY is present in cytotoxic granules of cytotoxic T lymphocytes and natural killer cells, and it has antimicrobial activity against M. tuberculosis and other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulysin
Applications Flow Cytometry
Classification Monoclonal
Clone eBioDH2 (DH2)
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene GNLY
Gene Accession No. P22749
Gene Alias D2S69E; GNLY; Granulysin; LAG2; LAG-2; lymphocyte-activation gene 2; Lymphokine; lymphokine LAG-2; Lymphokine NKG5519; NKG5; Protein NKG5; T-cell activation protein 519; TLA519; T-lymphocyte activation gene 519
Gene Symbols GNLY
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10578
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.