Learn More
Invitrogen™ Granzyme B Monoclonal Antibody (GB12), PE

Description
According to the literature this antibody recognizes granzyme B1 . Each lot is tested by flow cytometry using peripheral blood leukocytes. Based on this testing it is recommended that 5 μL of antibody be used per 1 x 10^6 cells. For R-PE conjugates excitation 488 nm/peak emission 575 nm. Matching isotype control is available: Ms IgG1 R-PE #MG104.
Specifications
Specifications
| Antigen | Granzyme B |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | GB12 |
| Concentration | 0.15 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 10% sucrose, 4mg/mL BSA and 0.1% sodium azide |
| Gene | GZMB |
| Gene Accession No. | P10144 |
| Gene Alias | AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.