Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granzyme M Monoclonal Antibody (4B2G4), eFluor™ 660, eBioscience™
GREENER_CHOICE

Catalog No. p-7173526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-4542 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-4542 Supplier Invitrogen™ Supplier No. 50977442
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Granzyme M is one of five granzyme serine proteases that have been identified in humans. These proteins are expressed in the granules of NK cells and cytotoxic T cells, and are critical for the induction of target cell apoptosis through the cleavage of intracellular substrates. Although not as well characterized as some of the other family members, Granzyme M has been found to exhibit some distinct functions. These include the non-cytotoxic inhibition of cytomegalovirus replication, induction of apoptosis in tumor cells, and contribution to the TLR4-induced inflammatory process. Applications Reported: This 4B2G4 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 4B2G4 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor 660 is a replacement for Alexa Fluor 647. eFluor 660 emits at...

Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme M
Applications Flow Cytometry
Classification Monoclonal
Clone 4B2G4
Concentration 5 μL/Test
Conjugate eFluor 660
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene GZMM
Gene Accession No. P51124
Gene Alias granzyme M; granzyme M (lymphocyte met-ase 1); GZMM; Hu-Met-1; Lmet1; lymphoctye Met-ase 1; lymphocyte met-ase 1; MET1; Met-1 serine protease; Met-ase; MMET-1; natural killer cell granular protease; RNK-Met-1
Gene Symbols GZMM
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3004
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.