Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme Oligo, Mouse Anti-Human, Clone: GB11, BD AbSeq™

Catalog No. BDB633793 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB633793 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB633793 Supplier BD Biosciences Supplier No. 940517
Add to Cart
Add to Cart

Mouse Oligoclonal Antibody

Granzyme Oligo Oligoclonal antibody specifically detects Granzyme Oligo in Human (Tested in Development) samples. It is validated for Single Cell 3'Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme Oligo
Applications Single Cell 3' Sequencing
Classification Oligoclonal
Clone GB11
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias GZMB; Granzyme-2; CCPI; CGL1; CSPB; CTLA1; CTSGL1; GRB; HLP; SECT
Host Species Mouse
Immunogen Human Granzyme B
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3002
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.