Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grape Flavor, Artificial, Concentrate, Spectrum™ Chemical
SDP

Catalog No. 18603046 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-046 500 mL Amber Glass Bottle
18-603-044 25 mL Amber Glass Bottle
18-603-045 100 mL Amber Glass Bottle
18-603-047 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-046 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G1427500ML
Add to Cart
Add to Cart

G1427

Spectrum™ Chemical Grape Flavor, Artificial, Concentrate

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.