Learn More
Sigma Organic Chemistry GRAPHENE OXIDE CARBOXYLIC ACID ENRICHED 2 MG/ML DISPERSION IN WATER | 50ML
Supplier: Sigma Organic Chemistry 79554250ML

GRAPHENE OXIDE CARBOXYLIC ACID ENRICHED 2 MG/ML DISPERSION IN WATER, 50ML
About This Item:
Linear Formula: CxHyOz
UNSPSC Code: 12352200
Storage: ROOM TEMPERATURE
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.