Learn More
Strem, An Ascensus Company GRAPHENE OXIDE (0.8-1.2NM THIC
SDP
Supplier:  Strem, An Ascensus Company 060323250MG
Encompass_Preferred
Graphene oxide (0.8-1.2nm thick x 1-15 microns wide, made by the Staudenmaier Method)
Catalog No. NC2160592
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.