Learn More
Perkin Elmer US LLC Graphite Front Ferrules - 0.125 in, Pkg. 1
Supplier:  Perkin Elmer US LLC 09920593
Encompass_Preferred
Of the three ferrule types available, graphite is the softest, making it less damaging to capillary columns when overtightened. Graphite seals easily and does not stick to glass columns.
Catalog No. 50-211-2711
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.