Learn More
Graphite powder, synthetic, conducting grade, -325 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
Description
Graphite powder is basically used for casting polishing purposes and molding. Natural graphite is mostly used for refractories, batteries, steel making, expanded graphite, brake linings, foundry facings and lubricants. It is also used in pigments, anodes arc-lamp carbons, coating cathode ray tubes and moderator in nuclear piles. Graphite (carbon) fiber and carbon nanotubes are also used in carbon fiber reinforced plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Lubricants, pigments, anodes arc-lamp carbons, coating cathode ray tubes, moderator in nuclear piles
Specifications
Specifications
| Color | Black |
| Mesh Size | −325 Mesh |
| Odor | Odorless |
| Quantity | 227 g |
| UN Number | UN1325 |
| Solubility Information | Insoluble in water as well as other organic solvents. |
| Percent Purity | 99.9995% |
| Grade | UCP-2 grade, ultra 'F' purity |
| Physical Form | Powder |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H228-H319-H335 |
| Hazard Statement | GHS H Statement H228-H319-H335 Flammable solid. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 231-955-3 |
| RTECSNumber | MD9659600 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.