Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan Scientific and Medical™ Graphite Sealing Ring for PerkinElmer Liner
SDP

Catalog No. 0726522 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
For use with 4 mm OD liners
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
07-265-22 For use with 4 mm OD liners
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-265-22 Supplier Trajan Scientific and Medical™ Supplier No. 0726522

missing translation for 'validMainframeMsg'

Made up of graphite and used as sealing rings for PerkinElmer GC.

  • Suitable for PerkinElmer GC
  • Temperature stable up to 450°C

Specifications

For Use With (Equipment) For use with 4 mm OD liners
Material Graphite
Quantity 10/Pk.
Product Type Graphite Sealing Ring for PerkinElmer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.