Learn More
Invitrogen™ GRASP1 Polyclonal Antibody

Description
Immunogen sequence: KESSAVPARS LSSSPQAQPP RPAELSDEEV AELFQRLAET QQEKWMLEEK VKHLEVSSAS MAEDLCRKSA IIETYVMDSR IDVSVAAGHT DRSGLGSVLR DLVKPGDENL REMNK Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
Specifications
| Antigen | GRASP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GRIPAP1 |
| Gene Accession No. | Q4V328 |
| Gene Alias | 30kDa C-terminus form; AI854681; DXImx47e; Grasp1; GRASP-1; GRASP-1 C-terminal chain; GRIP1; GRIP-1; GRIP1 associated protein 1; GRIP1-associated protein 1; Gripap1; GRIP-associated protein 1; HCMV-interacting protein; Kiaa1167; mKIAA1167; MPMGp800B12492Q3; Sfc10 |
| Gene Symbols | GRIPAP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.