Learn More
Sklar Instruments Merit(TM) Graves Vaginal Speculum, Sklar , Straight, Stainless Steel, Reusable. EA.
Supplier: Sklar Instruments 98303
Merit(TM) Graves Vaginal Speculum, Sklar, Straight, Stainless Steel, Reusable. The Merit(TM) Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient. This product is a small sized speculum with 3 in. X 3/4 in. blades. EA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.