Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Gravex™ Safety Eyewear

Built-in nose piece secures the glass and prevents slipping

$4.97 - $43.25

Specifications

Certifications/Compliance ANSI Z87.1
Frame Color Black
Weight (Metric) 25 g
Lens Material Polycarbonate
Frame Material Nylon
View More Specs

Products 4
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
19-155-868
View Documents
Pyramex™
SB8920S
Gray
Case of 12 for $43.25
Only null left
Add to Cart
 
19-155-869
View Documents
Pyramex™
SB8980S
Indoor/Outdoor, Mirror
Each for $4.97
Case of 12 EA for $56.50
Only null left
Add to Cart
 
19-155-866
View Documents
Pyramex™
SB8910S
Clear N/A N/A N/A
19-155-867
View Documents
Pyramex™
SB8910ST
Anti-fog Clear N/A N/A N/A
Description

Description

  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • 9.75 base curve lens provides full side protection without distracting seams or traditional type side shields
  • Specifications

    Specifications

    ANSI Z87.1
    25 g
    Nylon
    Black
    Polycarbonate
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.