Learn More
New Pig Gray Ham-O™ Absorbent Mat Roll in Dispenser Box
SDP
Catalog No. 18389161 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Gray Ham-O™ Absorbent Mat Roll in Dispenser Box
Catalog No.
18-389-161 Type: Gray Ham-O™ Absorbent Mat Roll in Dispenser Box
1 options
This item is not returnable. View return policy
Catalog No. 18-389-161 Supplier New Pig Supplier No. MAT1000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Brings the fun to leak, drip and spill cleanup! Rolls are perforated for tearing off just what you need and in a dispenser box for easy storing.
  • Narrow heavyweight rolls are the perfect size for lining shelves and ledges along machines and work best for soaking up heavy leaks and drips; dispenser box fits neatly on shelves or carts
  • PIG-Printed Camouflage Mats are a fun way to absorb liquid and hide grime
  • Absorbed leaks and drips blend right in to hamouflage top layer so mat appears clean
  • Walk all over it! Scuff-resistant hamouflage layer stands up to foot traffic in aisles and walkways; resists rips, tears and abrasions from tools when used as a liner
  • Use anywhere – line workbenches, counters and toolboxes, clean up oil and grease in service pits and more!
  • PIG Mat lasts 2x longer than competitive mats and won't rip, tear or fray even when saturated; liquids absorb quickly for faster, easier cleanups and safer workplaces
  • Easy-tear perforations let you tear off small or large pieces for less waste
  • Absorbs most common industrial liquids - oils, coolants, solvents, water and more
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending