Learn More
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (88%), Rat (86%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | GRB14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300 |
| Gene | GRB14 |
| Gene Accession No. | Q14449, Q9JLM9 |
| Gene Alias | AI505286; GRB14; GRB14 adapter protein; growth factor receptor bound protein 14; growth factor receptor-bound protein 14 |
| Gene Symbols | GRB14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
