Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRB2 Monoclonal Antibody (11-B11)
GREENER_CHOICE

Catalog No. PIMA534887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534887 Supplier Invitrogen™ Supplier No. MA534887
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 293, MCF-7, HepG2, rat brain tissue, human colon cancer tissue,.

GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2-Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 11-B11
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene GRB2
Gene Accession No. P62993
Gene Alias AA408164; abundant SRC homology; adapter protein GRB2; Adapter protein GRB2-A; ASH; ASH protein; EGFRBP-GRB2; epidermal growth factor receptor-binding protein GRB2; grb2; grb2.L; grb2-a; grb2-b; Grb3-3; growth factor receptor bound protein 2; growth factor receptor bound protein 2 L homeolog; growth factor receptor-bound protein 2; Growth factor receptor-bound protein 2-A; growth factor receptor-bound protein 3; HT027; hypothetical protein LOC535298; MST084; MSTP084; NCKAP2; OTTHUMP00000166097; OTTHUMP00000166098; protein Ash; SH2/SH3 adapter GRB2; SH2/SH3 adapter GRB2-A; SH2/SH3 adaptor Grb2; XELAEV_18044055mg
Gene Symbols GRB2
Host Species Mouse
Immunogen Recombinant protein within Human GRB2 aa 21-217.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2885
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.