Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA110033
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA110033 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA110033 Supplier Invitrogen™ Supplier No. PA110033
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes Grb2, a highly conserved approximately 26 kDa intracellular adaptor protein. The immunogen has 100% identity between man, cow, rat and mouse. Western Blot: Reducing conditions.

GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2-Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene GRB2
Gene Accession No. P62993
Gene Alias AA408164; abundant SRC homology; adapter protein GRB2; Adapter protein GRB2-A; ASH; ASH protein; EGFRBP-GRB2; epidermal growth factor receptor-binding protein GRB2; grb2; grb2.L; grb2-a; grb2-b; Grb3-3; growth factor receptor bound protein 2; growth factor receptor bound protein 2 L homeolog; growth factor receptor-bound protein 2; Growth factor receptor-bound protein 2-A; growth factor receptor-bound protein 3; HT027; hypothetical protein LOC535298; MST084; MSTP084; NCKAP2; OTTHUMP00000166097; OTTHUMP00000166098; protein Ash; SH2/SH3 adapter GRB2; SH2/SH3 adapter GRB2-A; SH2/SH3 adaptor Grb2; XELAEV_18044055mg
Gene Symbols GRB2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human Grb2 (amino acids 198-217).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2885
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.